Learn More
Bon Opus Biosciences Recombinant Human IL-36a -50ug
SDP
Supplier:  Bon Opus Biosciences CR6150UG
Encompass_Preferred
Recombinant Human Interleukin-36 Alpha is produced by our E.coli expression system and the target gene encoding Lys6-Phe158 is expressed. Expression system: E.coli. Accession# Q9UHA7. Formulation: Lyophilized from a 0.2 um filtered solution of 50mM Tris HCl, 150mM NaCl, pH8.0,1 mM EDTA,0.02%Tween 20. Bon Opus Cat# CR61
Catalog No. 50-208-0394