Learn More
Bon Opus Biosciences Recombinant Human Interleukin-7/IL-7 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP086500UG
Encompass_Preferred
Recombinant Human Interleukin-7 is produced by Human 293 Cells. The target gene encoding D26-H177 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# P13232. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP086
Catalog No. 50-208-0161