Learn More
Bon Opus Biosciences Recombinant Human Interleukin-2 Receptor Subunit Beta/IL2RB (N-Avi/His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP056500UG
Encompass_Preferred
Recombinant human interleukin-2 receptor subunit beta is produced by Human 293 Cells. The target gene encoding A27-T240 is expressed with a Avi/;6His tag at the N terminus. Expression system: Human 293 Cells. Accession# P14784. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP056
Catalog No. 50-208-0131