Learn More
Bon Opus Biosciences Recombinant Human Leydig Insulin-Like 3/INSL3 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP024500UG
Encompass_Preferred
Recombinant Human Insulin-like 3 is produced by our Mammalian expression system and the target gene encoding Leu21-Tyr131 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# P51460. Formulation: Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP024
Catalog No. 50-208-0107