Learn More
Bon Opus Biosciences Recombinant Human Insulin-Degrading Enzyme/IDE/Insulysin (C-6His)
SDP
Supplier:  Bon Opus Biosciences CA0550UG
Encompass_Preferred
Recombinant Human Insulin-Degrading Enzyme is produced by our Mammalian expression system and the target gene encoding Met42-Leu1019 is expressed with a 6His tag at the C-terminus. Bon Opus Cat. #CA05 Accession# P14735. Bon Opus Cat# CA05
Catalog No. 50-203-2623