Learn More
Bon Opus Biosciences Recombinant Human Interleukin-15/IL-15 500UG
SDP
Supplier:  Bon Opus Biosciences C016500UG
Encompass_Preferred
Recombinant Human Interleukin-15 is produced by our E.coli expression system and the target gene encoding Asn49-Ser162 is expressed. Accession# P40933. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.0. Bon Opus Cat# C016
Catalog No. 50-217-2716