Learn More
Bon Opus Biosciences Recombinant Human Interleukin-18/IL-18/IL-1F4 (C-6His) 10ug
SDP
Supplier:  Bon Opus Biosciences CX7210UG
Encompass_Preferred
Recombinant Human Interleukin-18 is produced by our Mammalian expression system and the target gene encoding Tyr37-Asp193 is expressed with a 6His tag at the C-terminus. Accession# Q14116. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,pH7.4. Bon Opus Cat# CX72
Catalog No. 50-217-2835