Learn More
Bon Opus Biosciences Recombinant Human Interleukin-36γ/IL-36γ/IL-1F9 500ug
SDP
Supplier:  Bon Opus Biosciences CM77500UG
Encompass_Preferred
Recombinant Human Interleukin-36 gamma is produced by our E.coli expression system and the target gene encoding Ser18-Asp169 is expressed. Accession# Q9NZH8. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM Tris,100mM Nacl,0.1mM EDTA,pH8.0. Bon Opus Cat# CM77
Catalog No. 50-217-2823