Learn More
Bon Opus Biosciences Recombinant Human/Mouse/Rat Irisin (N-6His) -50ug
SDP
Catalog No. 502080310
Encompass_Preferred
Recombinant Human/Mouse/Rat Fibronectin type III domain-containing protein 5  is produced by our Mammalian expression system and the target gene encoding Asp32-Glu143 is expressed with a 6His tag atthe N-terminus. Expression system: Human Cells. Accession# Q8NAU1. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C00B
Catalog No. 50-208-0310 Supplier Bon Opus Biosciences Supplier No. C00B50UG
Recombinant Human/Mouse/Rat Fibronectin type III domain-containing protein 5  is produced by our Mammalian expression system and the target gene encoding Asp32-Glu143 is expressed with a 6His tag atthe N-terminus. Expression system: Human Cells. Accession# Q8NAU1. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C00B