Learn More
Bon Opus Biosciences Recombinant Human Kallikrein 7/KLK7/SCEE (C-6His 500ug
SDP
Supplier:  Bon Opus Biosciences C364500UG
Encompass_Preferred
Recombinant Human Kallikrein 7 is produced by our Mammalian expression system and the target gene encoding Glu23-His252 is expressed with a 6His tag at the C-terminus. Accession# P49862. Formulation: Supplied as a 0.2 μm filtered solution of 20mM HEPES, 150mM NaCl, pH 7.5. Bon Opus Cat# C364
Catalog No. 50-217-2737