Learn More
Bon Opus Biosciences Recombinant Human Prostate-Specific Antigen//PSA/KLK3 (Ala18-Pro261, C-6His) 10ug
SDP
Supplier:  Bon Opus Biosciences C36510UG
Encompass_Preferred
Recombinant Human Kallikrein 3 is produced by our Mammalian expression system and the target gene encoding Ala18-Pro261 is expressed with a 6His tag at the C-terminus. Accession# P07288. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM MES, 150mM NaCl, 2mM CaCl2, pH 6.0. Bon Opus Cat# C365
Catalog No. 50-217-2738