Learn More
Bon Opus Biosciences Recombinant Human KRAS4B(G12C, N-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CR6750UG
Encompass_Preferred
Recombinant Human GTPase Kras4B is produced by our E.coli expression system and the target gene encoding Met1-Lys169(Gly12Cys) is expressed with a 6His tag at the N-terminus. Expression system: E.coli. Accession# P01116-2. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR67
Catalog No. 50-208-0400