Learn More
Bon Opus Biosciences Recombinant Human LAIR1 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP002500UG
Encompass_Preferred
Recombinant Human Leukocyte-Associated Immunoglobulin-Like Receptor 1 is produced by human 293 cells. The target gene encoding Gln22-Tyr165 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q6GTX8. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PB, 15umM NaCl, pH 7.2. Bon Opus Cat# BP002
Catalog No. 50-208-0083