Learn More
Bon Opus Biosciences Recombinant Human LAIR2 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences C02C50UG
Encompass_Preferred
Recombinant Human Leukocyte-Associated Immunoglobulin-Like Receptor 2 is produced by our Mammalian expression system and the target gene encoding Gln22-Pro152 is expressed with a 6His tag at the C-terminus.  Expression system: Human Cells. Accession# Q6ISS4. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C02C
Catalog No. 50-208-0342