Learn More
Bon Opus Biosciences Recombinant Human Leukemia Inhibitory Factor/LIF -500ug
SDP
Supplier:  Bon Opus Biosciences BP166500UG
Encompass_Preferred
Recombinant Human Leukemia Inhibitory Factor is produced by our E.coli expression system and the target gene encoding Ser23-Phe202 is expressed. Expression system: E. coli. Accession# P15018. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 0.02% Tween 20, pH 7.4. Bon Opus Cat# BP166
Catalog No. 50-208-0264