Learn More
Bon Opus Biosciences Recombinant Human Leukocyte Ig-Like Receptor B1/LILRB1/ILT2/CD85j (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP161500UG
Encompass_Preferred
Recombinant Human Leukocyte Immunoglobulin-Like Receptor Subfamily B Member 1 is produced by our Mammalian expression system and the target gene encoding Gly24-His458 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q8NHL6. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP161
Catalog No. 50-208-0256