Learn More
Bon Opus Biosciences Recombinant Human LILRB4 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW5050UG
Encompass_Preferred
Recombinant Human Leukocyte Immunoglobulin-like Receptor Subfamily B Member 4 is produced by our Mammalian expression system and the target gene encoding Gln22-Glu259 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q8NHJ6. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW50
Catalog No. 50-208-0433