Learn More
Bon Opus Biosciences Recombinant Human LILRB4 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW4950UG
Encompass_Preferred
Recombinant Human Leukocyte Immunoglobulin-like Receptor Subfamily B Member 4 is produced by our Mammalian expression system and the target gene encoding Gln22-Glu259 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q8NHJ6. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW49
Catalog No. 50-208-0432