Learn More
Cell Systems Corporation Primary Human Liver Sinusoidal Microvascular Endothelial Cells (ACBRI 566 V)
SDP
Catalog No. NC1876690
Encompass_Preferred
Primary Human Liver Sinusoidal Microvascular Endothelial Cells (ACBRI 566 V) were initiated from normal human liver tissue. Unlike cells from other providers, Cell Systems primary cells are isolated without antibody labels for enhanced biological relevance. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.
Catalog No. NC1876690 Supplier Cell Systems Corporation Supplier No. ACBRI 566 V
Add to Cart
Add to Cart
Primary Human Liver Sinusoidal Microvascular Endothelial Cells (ACBRI 566 V) were initiated from normal human liver tissue. Unlike cells from other providers, Cell Systems primary cells are isolated without antibody labels for enhanced biological relevance. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.