Learn More
Bon Opus Biosciences Recombinant Human LYVE-1 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP17450UG
Encompass_Preferred
Recombinant Human Lymphatic Vessel Endothelial Hyaluronic Acid Receptor 1 is produced by our Mammalian expression system and the target gene encoding Leu20-Thr238 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q9Y5Y7. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM Tris-Citrate,1 50mM NaCl, pH 7.0. Bon Opus Cat# BP174
Catalog No. 50-208-0277