Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokine 30-Plex Human Panel

Catalog No. lhc6003m
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LHC6003M 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LHC6003M Supplier Invitrogen™ Supplier No. LHC6003M
Only null left
Add to Cart
Add to Cart
Request a Quote

Includes

Contains premixed antibody coated capture beads, standard, premixed detector antibody, diluents, SAV concentrate, buffers, wash solution, filter plate, flat bottom plate, complete protocol and lot-specific technical data sheet.

Specifically designed for quantifying human cytokines, chemokines and growth factors in serum, plasma, and tissue culture supernatant

By measuring 30 analytes simultaneously, the assay provides more data from each sample, saving costs and time. This assay may be run alone or in combination with other select Luminex singleplex kits. The panel employs magnetic beads, facilitating automation, decreasing hands-on time, and increasing throughput and precision. Suitable for use with Luminex™ 100/200, FLEXMAP 3D™ and MAGPIX™ systems.

Comprehensive Human Analyte Panel
The Human Cytokine Magnetic 30-Plex Panel for the Luminex platform provides reagents for the accurate, reproducible, and sensitive quantitation of human proteins including:

CytoKines: G-CSF, GM-CSF, IFN-α, IFN-γ, IL-1β, IL-1RA , IL-2, IL-2R, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p40/p70), IL-13, IL-15, IL-17, TNF-α

Chemokines: Eotaxin, IP-10, MCP-1, MIG, MIP-1α, MIP-1β, RANTES

Growth factors: EGF, FGF-basic, HGF, VEGF


Luminex xMAP™ Technology—Effective and Proven Analytical Tool
The Human Cytokine Magnetic 30-Plex Panel is based on xMAP technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex assays. Magnetic microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. At the completion of the sandwich immunoassay, beads can be read using the Luminex 200 or FLEXMAP 3D detection systems, instruments based on flow cytometry, two lasers, real-time digital signal processing, and xPONENT™ software to distinguish bead color (analyte) and assay signal strength (PE) fluorescence intensity.

MAGPIX System—Brings the Power of Multiplex to Every Lab
The MAGPIX system may also be used for sample analysis. This instrument utilizes xMAP; technology and is built on light-emitting diodes (LEDs) for excitation, a CCD camera for bead and analyte detection and high-speed digital signal processors. The MAGPIX system is more compact and robust, providing a very cost effective magnetic bead-based multiplexing tool. Simple, out of the box set-up allows researchers to rapidly perform singleplex or multiplex assays of up to 50 targets in single biological samples. The MAGPIX system is easy to use—ideal for both new and experienced users alike.

Magnetic Beads Let You Do More with Your Time
Magnetic bead-based assays use Luminex MagPlex magnetic microsphere. All other assay components are the same as the equivalent Luminex polystyrene bead-based assays providing identical quality and consistency performance. MagPlex technology takes advantage of magnetic properties to simplify assay wash steps and maximize uniformity of results. Magnetic bead-based assays also enable automation, decrease hands-on time, and increase throughput and precision. Easy to use protocols allow you to get your results in as little as 3.5 hours. In addition, these assays are compatible with both vacuum and magnetic wash stations and work well with the MAGPIX system as well as other Luminex xMAP platforms.

More Validation for Confidence in Your Results
The Human Cytokine Magnetic 30-Plex Panel has been validated using the same rigorous criteria that distinguishes Luminex assays. Each kit supplied with a product insert that outlines the specifications for each marker in the assay.

  • Superior performance: accurate, reproducible, and sensitive quantitation of multiple proteins
  • High quality: fully qualified antibodies permit excellent specificity and sensitivity
  • Fast and easy protocols: perform your multiplex assay and analyze your data typically in less than one day

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 100 tests
Species Human
Combinability Combinable
Detection Method Fluorescence
Conjugate R-PE
Bead Type See product documentation
Assay Sensitivity See Certificate of Analysis
Assay Range See product documentation
Format Preconfigured Panel
For Use With (Equipment) Luminex™ Instruments
Product Line Novex
Protein Cytokine
Protein Subtype Cytokines & Receptors
Sample Type Serum, Plasma, Cell Culture Supernatants
Content And Storage Contains premixed antibody coated capture beads, standard, premixed detector antibody, diluents, SAV concentrate, buffers, wash solution, flat bottom plate, complete protocol and lot-specific technical data sheet. Store at 2°C to 8°C.
Shipping Condition Wet Ice
Research Area Immunology, Cytokines
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.