Learn More
Bon Opus Biosciences Recombinant Human Mannan-Binding Lectin Serine Protease 1/MASP1 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences BP219500UG
Encompass_Preferred
Recombinant Human Mannan-Binding Lectin Serine Protease 1 is produced by our Mammalian expression system and the target gene encoding His20-Arg728 is expressed with a 6His tag at the C-terminus. Accession# P48740. Formulation: Supplied as a 0.2 μm filtered solution of 20mM Tris,200mM NaCl,10%Glycerol,pH8.0. Bon Opus Cat# C655
Catalog No. 50-217-2753