Learn More
Bon Opus Biosciences Recombinant Human Methyl-CpG-Binding Protein 2/MECP2 (C-6His) 500ug
Supplier: Bon Opus Biosciences CI08500UG
Recombinant Human MECP2 is produced by our Mammalian expression system and the target gene encoding Met1-Ser486 is expressed with a 6His tag at the C-terminus. Accession# P51608. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM Tris,150mM NaCl,pH 8.0. Bon Opus Cat# CI08