Learn More
Bon Opus Biosciences Recombinant Human Trans-2-Enoyl-CoA Reductase Mitochondrial/MECR (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C9171MG
Encompass_Preferred
Recombinant Human Trans-2-Enoyl-CoA Reductase is produced by our Mammalian expression system and the target gene encoding Pro54-Met373 is expressed with a 6His tag at the C-terminus. Accession# Q9BV79. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# C917
Catalog No. 50-217-2762