Learn More
Bon Opus Biosciences Recombinant Human Mesothelin/MSLN (C-His) - Source: Human 293 Cells
SDP
Catalog No. NC2733674
Encompass_Preferred
Recombinant Human Mesothelin is produced by our Mammalian expression system and the target gene encoding Glu296-Ser598 is expressed with a 6His tag at the C-terminus. Accession#AAH09272.1. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4
Catalog No. NC2733674 Supplier Bon Opus Biosciences Supplier No. BP24650UG
Recombinant Human Mesothelin is produced by our Mammalian expression system and the target gene encoding Glu296-Ser598 is expressed with a 6His tag at the C-terminus. Accession#AAH09272.1. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4