Learn More
Bon Opus Biosciences Recombinant Human Myelin Oligodendrocyte Glycoprotein/MOG (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP169500UG
Encompass_Preferred
Recombinant Human Myelin Oligodendrocyte Glycoprotein is produced by our Mammalian expression system and the target gene encoding Gly30-Gly154 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q16653. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP169
Catalog No. 50-208-0268