Learn More
Bon Opus Biosciences Recombinant Human Mortality Factor 4-Like Protein 2/MORF4L2/MRGX (C-6His) - Source: E. coli
SDP
Catalog No. NC2733766
Encompass_Preferred
Recombinant Human Mortality Factor 4-Like Protein 2 is produced by our E.coli expression system and the target gene encoding Met1-Leu288 is expressed with a His tag at the C-terminus. Accession#Q15014. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4.
Catalog No. NC2733766 Supplier Bon Opus Biosciences Supplier No. BP29950UG
Recombinant Human Mortality Factor 4-Like Protein 2 is produced by our E.coli expression system and the target gene encoding Met1-Leu288 is expressed with a His tag at the C-terminus. Accession#Q15014. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4.