Learn More
Bon Opus Biosciences Recombinant Human Mortality Factor 4-Like Protein 2/MORF4L2/MRGX (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CE43500UG
Encompass_Preferred
Recombinant Human Mortality Factor 4-Like Protein 2 is produced by our E.coli expression system and the target gene encoding Met1-Leu288 is expressed with a 6His tag at the C-terminus. Accession# Q15014. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CE43
Catalog No. 50-217-2779