Learn More
Lonza Walkersville Hu Naive CD8/CD45RA T-Cells
Catalog No. 5000496294
Encompass
Leukapheresed Na ve CD8 T cells are isolated using a two step isolation process CD8 T Cells are first negatively selected from MNC using an indirect immunomagnetic CD8 labeling system CD45RO microbeads are then used to deplete the CD45RO population leaving a highly enriched CD4 CD45RA Na ve T Cell population Cells are cryopreserved in Lonza Cryo Media and collected in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols Manufactured by AllCells
Catalog No. 50-004-96294 Supplier Lonza Walkersville Supplier No. 4W302
Leukapheresed Na ve CD8 T cells are isolated using a two step isolation process CD8 T Cells are first negatively selected from MNC using an indirect immunomagnetic CD8 labeling system CD45RO microbeads are then used to deplete the CD45RO population leaving a highly enriched CD4 CD45RA Na ve T Cell population Cells are cryopreserved in Lonza Cryo Media and collected in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using AO/PI Cells are collected from healthy donors following IRB protocols Manufactured by AllCells