Learn More
Bon Opus Biosciences Recombinant Human Nectin-4 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW9550UG
Encompass_Preferred
Recombinant Human Poliovirus Receptor-Related Protein  4 is produced by our Mammalian expression system and the target gene encoding Gly31-Val351 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q96NY8. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CW95
Catalog No. 50-208-0472