Learn More
Bon Opus Biosciences Recombinant Human NKG2A/CD94 Heterodimer 1mg
SDP
Supplier:  Bon Opus Biosciences BP0011MG
Encompass_Preferred
Recombinant Human NKG2A & CD94 Heterodimer Protein is produced by human 293 cells. The target gene encoding Arg100-Leu233 & Ser34-Ile179 is expressed with a 8His & Flag tag at the N-terminus. Accession# P26715 & Q13241. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4 Bon Opus Cat# BP001
Catalog No. 50-217-2602