Learn More
Bon Opus Biosciences Recombinant Human Noggin/NOG 500ug
SDP
Supplier:  Bon Opus Biosciences CB89500UG
Encompass_Preferred
Recombinant Human Noggin is produced by our Mammalian expression system and the target gene encoding Gln28-Cys232 is expressed. Accession# Q13253. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,500mM NaCl,2mM EDTA,pH7.4. Bon Opus Cat# CB89
Catalog No. 50-217-2773