Learn More
Abcam Human NPTX2 ELISA Kit - Intracellular, 1 x 96 Tests
Supplier:  Abcam AB3148471X96TESTS
Encompass_Preferred
Human NPTX2 ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify Human NPTX2 with a sensitivity of 12.535 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2748834