Learn More
Bon Opus Biosciences Recombinant Human Neuropilin-1/NRP1 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP183500UG
Encompass_Preferred
Recombinant Human Neuropilin-1 is produced by our Mammalian expression system and the target gene encoding Phe22-Lys644 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# O14786. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 5% Trehalose, 0.01%Tween 80, pH7.4. Bon Opus Cat# BP183
Catalog No. 50-208-0292