Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCEGEN BIOTECH HU ORBITAL FIBROBLAST MED KIT

Catalog No. NC3245911
Encompass

  • Cell culture medium kit
  • Primary cell culture use
  • Target cells: human orbital fibroblasts
  • Compatible with human species
  • Applications in Graves' ophthalmopathy research
  • Supports spindle-shaped cell morphology
  • Key markers supported: vimentin, fibronectin
  • Facilitates extracellular matrix production
  • Helps in tissue repair processes
  • Enhances tissue remodeling capabilities
  • Promotes high cell viability
  • Source of cells: human orbital connective tissue

Catalog No. NC3245911 Supplier ACCEGEN BIOTECH Supplier No. ABMTM077G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Cell culture medium kit
  • Primary cell culture use
  • Target cells: human orbital fibroblasts
  • Compatible with human species
  • Applications in Graves' ophthalmopathy research
  • Supports spindle-shaped cell morphology
  • Key markers supported: vimentin, fibronectin
  • Facilitates extracellular matrix production
  • Helps in tissue repair processes
  • Enhances tissue remodeling capabilities
  • Promotes high cell viability
  • Source of cells: human orbital connective tissue

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.