Learn More
Sciencell Research Laboratories Human Ovarian Surface Epithelial Cells, Catalog #7310, Frozen, >5 x 10^5 cells in 1 ml
Catalog No. NC3718242
Encompass
Human ovarian surface epithelial cells (HOSEpiC) are primary cells derived from human ovarian tissue and are vital for various research in ovarian physiology. Ideal for studying follicular dynamics, hormone expression, and the underlying mechanisms in ovarian cancers, these cells provide significant insights into cell survival and transformation processes.
  • Isolated from human ovary
  • Cryopreserved at early passage
  • Delivered in frozen state
  • Contains >5 x 10^5 cells in 1 ml
  • Characterization markers include Cytokeratin-18 and/or Cytokeratin-19
  • Negative pathogen testing for HIV-1, HBV, HCV, and others
  • Guaranteed expansion of 5 population doublings
  • Recommended medium is Ovarian Epithelial Cell Medium (OEpiCM)
  • For research use only
Catalog No. NC3718242 Supplier Sciencell Research Laboratories Supplier No. 7310
Human ovarian surface epithelial cells (HOSEpiC) are primary cells derived from human ovarian tissue and are vital for various research in ovarian physiology. Ideal for studying follicular dynamics, hormone expression, and the underlying mechanisms in ovarian cancers, these cells provide significant insights into cell survival and transformation processes.
  • Isolated from human ovary
  • Cryopreserved at early passage
  • Delivered in frozen state
  • Contains >5 x 10^5 cells in 1 ml
  • Characterization markers include Cytokeratin-18 and/or Cytokeratin-19
  • Negative pathogen testing for HIV-1, HBV, HCV, and others
  • Guaranteed expansion of 5 population doublings
  • Recommended medium is Ovarian Epithelial Cell Medium (OEpiCM)
  • For research use only