Learn More
Bon Opus Biosciences Recombinant Human OX40 Ligand/TNFSF4/OX40L (N-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CJ45500UG
Encompass_Preferred
Recombinant Human OX40 Ligand is produced by our Mammalian expression system and the target gene encoding Gln51-Leu183 is expressed with a 6His tag at the N-terminus. Accession# P23510. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl, pH7.4. Bon Opus Cat# CJ45
Catalog No. 50-217-2813