Learn More
Bon Opus Biosciences Recombinant Human Cellular Tumor Antigen p53 (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP055500UG
Encompass_Preferred
Recombinant human cellular tumor antigen p53 is produced by E.coli. The target gene encoding M1-D393 is expressed with a 6His tag at the N terminus. Expression system: E. coli. Accession# P04637. Formulation: Supplied as a 0.2 um filtered solution of 2umM Tris,30umM NaCl ,5% glycerol ,umM DTT, 0.0umM ZnCl2, pH8.0. Bon Opus Cat# BP055
Catalog No. 50-208-0130