Learn More
Lonza Walkersville Hu PB CD3 Pan T-Cells 25M
Catalog No. 5000496334
Encompass
Pan T Cells (CD3 ) are negatively selected from mononuclear cells obtained from leukaparesed blood and using an indirect immunomagnetic labeling system Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using hemacytometer (trypan blue method) Cells are collected from healthy donors following IRB protocols
Catalog No. 50-004-96334 Supplier Lonza Walkersville Supplier No. 2W350
Pan T Cells (CD3 ) are negatively selected from mononuclear cells obtained from leukaparesed blood and using an indirect immunomagnetic labeling system Cells are cryopreserved in Lonza Cryo Media in a cryovial Cell purity is determined post thaw using a FACS assay Count and viability is determined using hemacytometer (trypan blue method) Cells are collected from healthy donors following IRB protocols