Learn More
Lonza Walkersville Hu PB CD8 T-Cells 10M
Supplier:  Lonza Walkersville 2W300
Encompass
Peripheral blood CD8 T cells also referred to as cytotoxic T cells are critical components of the adaptive immune response CD8 is a transmembrane glycoprotein which serves as a co-receptor for TCR (T Cell Receptor) to recognize antigens on major histocompatibility complex (MHC) Once bound CD8 T cells kill the infected cell by secreting cytotoxic granules containing granzymes and perforin resulting in apoptosis of the target cell Cells You Can Count on to PerformLonza CD8 T Cells are isolated from peripheral blood of healthy donors using negative immunomagnetic separation Cryopreserved CD8 T cells are guaranteed 85% pure for CD8 T cells as assessed by flow cytometry They are guaranteed to contain 10 million viable cells per vial HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Cells are available from multiple donors
Catalog No. 50-004-96228