Learn More
HUMANCELLS BIOSCIENCES HU PBMC FR 50MM BL MONO CLS
Catalog No. NC2110997
Encompass
Human PBMC Fresh 50 MillionHuman Peripheral Blood Mononuclear Cells (PBMCs) are blood cells with a single round nucleus. These cells include lymphocytes (T, B and NK cells), monocytes and dendritic cells. PBMCs are parts of the immune system which are critical to cell-mediated and humoral immunity.
This item is not returnable. View return policy
Catalog No. NC2110997 Supplier HUMANCELLS BIOSCIENCES Supplier No. PBMCF50M
Human PBMC Fresh 50 MillionHuman Peripheral Blood Mononuclear Cells (PBMCs) are blood cells with a single round nucleus. These cells include lymphocytes (T, B and NK cells), monocytes and dendritic cells. PBMCs are parts of the immune system which are critical to cell-mediated and humoral immunity.