Learn More
Bon Opus Biosciences Recombinant Human Peptidase D/PEPD 500ug
Supplier: Bon Opus Biosciences CG88500UG
Recombinant Human Peptidase D is produced by our E.coli expression system and the target gene encoding Ala2-Lys493 is expressed. Accession# P12955. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4 Bon Opus Cat# CG88