Learn More
Abcam Human Perforin ELISA Kit (PRF1), 2 x 96 Tests
Supplier:  Abcam AB460682X96TESTS
Encompass_Preferred
Human Perforin ELISA Kit (PRF1) is a Sandwich (quantitative) ELISA kit for the measurement of Human Perforin (PRF1) in Human in Cell culture supernatant samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1978001