Learn More
Bon Opus Biosciences Recombinant Human Fructose-2,6-Bisphosphatase 1/PFKFB1/PFK/FBPase 1 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CI45500UG
Encompass_Preferred
Recombinant Human FBPase 1 is produced by our Mammalian expression system and the target gene encoding Ser2-Tyr471 is expressed with a 6His tag at the C-terminus. Accession# P16118. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4 Bon Opus Cat# CI45
Catalog No. 50-217-2802