Learn More
Bon Opus Biosciences Recombinant Human 6-Phosphogluconate Dehydrogenase, Decarboxylating/PGD (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CI74500UG
Encompass_Preferred
Recombinant Human 6PGD is produced by our Mammalian expression system and the target gene encoding Met1-Ala483 is expressed with a 6His tag at the C-terminus. Accession# P52209. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CI74
Catalog No. 50-217-2805