Learn More
Bon Opus Biosciences Recombinant Human Polymeric Immunoglobulin Receptor/PIgR (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP074500UG
Encompass_Preferred
Recombinant Human Polymeric Immunoglobulin Receptor is produced by Human 293 Cells. The target gene encoding K19-R638 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P01833. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.5. Bon Opus Cat# BP074
Catalog No. 50-208-0149