Learn More
Bon Opus Biosciences Recombinant Human Prefoldin Subunit 4/PFDN4 (N-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CE13500UG
Encompass_Preferred
Recombinant Human Prefoldin Subunit 4 is produced by our E.coli expression system and the target gene encoding Met1-Ser134 is expressed with a 6His tag at the N-terminus. Accession# Q9NQP4. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# CE13
Catalog No. 50-217-2776