Learn More
Bon Opus Biosciences Recombinant Human PRL -50ug
SDP
Supplier:  Bon Opus Biosciences C73650UG
Encompass_Preferred
Recombinant Human Prolactin is produced by our E.coli expression system and the target gene encoding Leu29-Cys227 is expressed.  Expression system: E.coli. Accession# P01236. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM Nacl,1mM EDTA,pH7.4. Bon Opus Cat# C736
Catalog No. 50-208-0362