Learn More
Bon Opus Biosciences Recombinant Human Prion-Like Protein Doppel/PRND (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CJ221MG
Encompass_Preferred
Recombinant Human Prion-like protein doppel is produced by our Mammalian expression system and the target gene encoding Arg27-Gly152 is expressed with a 6His tag at the C-terminus. Accession# Q9UKY0. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4 Bon Opus Cat# CJ22
Catalog No. 50-217-2810