Learn More
Bon Opus Biosciences Recombinant Human Pro-Brain-Derived Neurotrophic Factor/pro-BDNF 1mg
SDP
Supplier:  Bon Opus Biosciences C0771MG
Encompass_Preferred
Recombinant Human pro-Brain-Derived Neurotrophic Factor is produced by our E.coli expression system and the target gene encoding Ala19-Arg247 is expressed. Accession# P23560. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 250mM NaCl, pH 7.2. Bon Opus Cat# C077
Catalog No. 50-217-2722